Under-the-Counter Acid Cabinets, SciMatCo
Fournisseur: Scientific Materials
Certificats
A propos de cet article
Constructed of 2.5 cm (1") thick plywood, to be rust and corrosion proof
- Finished with chemical resistant epoxy paint
- Cabinets include padlock and two keys for security
Note: Dimensions include wood edge to wood edge height measurement of 87.6 cm (34½") without leveler.
Cabinets are made to fit under existing fume hoods or can be used as stand alone units. Cabinets feature a fixed position shelf, 5.1 cm (2") liquid-tight trough to contain spillage, and a wooden hinge that eliminates deterioration. Two styles are available. Partially lined with polypropylene on shelf and floor or fully lined with polypropylene on all interior walls, shelf, floor and top, except inside door surface.
Color: Blue
Customers Who Bought This Also Bought