Qorpak® Black Phenolic Screw Caps, Pulp/Vinyl Liner
Fournisseur: Qorpak
Certificats
A propos de cet article
Phenolic caps with pulp and vinyl liners are perfect for general use
- Closure is made of phenolic resin
- Liner is made of vinyl coating applied to HDPE coated paper laminated to pulpboard
This general purpose liner is commonly used for food, beverage, medical and chemical applications packed at less than 120° F., including mild acids, alkalis, solvents, alcohols, oils and aqueous products. This closure is not recommended for active hydrocarbons or bleaches.
Customers Who Bought This Also Bought