KIMBLE® DUALL® Tissue Grinders, PTFE Pestles and Glass Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Tissue grinders greatly improve grinding efficiency by combining both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate
- Construction is strong enough to allow the pestle to be motor-driven
- Pestles are designed to be used with 788000 stirrer adapters
- Replacement components are available and completely interchangeable
- Clearance between pestles and tubes is 0.004” to 0.006”
This PTFE pestle model is ideal for soft tissue such as brain or liver.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Special clearance pestles are available upon request; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought