SX-80 SensaTemp® Sodr-X-Tractor™ Desoldering Handpiece, PACE®
Fournisseur: Pace
Certificats
A propos de cet article
Features an improved design with a disposable solder collection chamber and quick-change heating element
Handpiece uses Endura™ desoldering tips. These nonclogging tips require less maintenance and cleaning.
Thru-hole desoldering, thermo-drive desoldering, flo desoldering, PCMCIA fine pitch TSOP removal, and small PQFP removal are possible with this unit. For use with SensaTemp® stations.
Customers Who Bought This Also Bought