Pallflex® Tissuquartz™ Filters
Fournisseur: Cytiva
Certificats
A propos de cet article
Pallflex Tissuquartz filters are 100% pure quartz filters (with no binder) for air monitoring applications. They feature high flow rate and filtration efficiency.
- Pure quartz
- High flow rate
- Interference-free results
Obtain highly sensitive and interference-free results due to their reduced residual ion and organics content.
Tissuquartz filters can withstand high operating temperature. This feature makes them durable and ideal for analysis of acidic gas, stack sampling, and other harsh testing environments.
For even lower level of trace organics, choose the popular heat-treated grade 2500 QAT-UP.
Ordering information: Filter media is pure quartz, no binder.
Spécifications
- Catalog No:
- CA28150-907
- CA28150-903
- CA29301-690
- CA28150-901
- CA28150-899
- CA76339-872
- CA21431-416
- 76411-294
- CA76411-292
- CA28150-905
- CA28150-897
- Max. operating temperature:
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- 1093 °C (2000 °F); air
- pH in boiled water extract:
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- 6.5 - 7.5
- Typical aerosol retention:
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- 99.9% (DOP)
- Typical air flow rate:
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm² at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- 73 L/min/cm2 at 0.7 bar (70 kPa, 10 psi)
- Typical filter weight:
- 5.8 mg/cm2
- 5.8 mg/cm2
- 5.8 mg/cm2
- 5.8 mg/cm2
- 5.8 mg/cm2
- 5.8 mg/cm2
- 5.8 mg/cm2
- 5.8 mg/cm2
- 5.8 mg/cm²
- 5.8 mg/cm2
- 5.8 mg/cm2
- Typical thickness:
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- 432 µm (17 mils)
- Typical water flow rate:
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm² at 0.35 bar (35 kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)
- 220 mL/min/cm2 at 0.35 bar (35kPa, 5 psi)