Nalgene® Rapid-Flow™ Sterilization Filter Units, Cellulose Nitrate Membrane, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
For easy, fast, sterile filtration of 150 to 1000 ml volumes.
- Instantly ready for biological filtration, cold sterilization, or cleanup of particulated solutions or buffers
- Components separate easily for pouring
- No need to transfer sterile filtrate to another container
- Graduations on uppers and receiver
- Non-toxic, triton-free, preassembled and radiation sterilized
Nalgene 1¹/₂-turn threaded screw closure guarantees leakproof seal and eliminates pH shift in receiver bottles. Receiver bottles can be used to store filtrate. Filter cup is printed with catalog number, lot number, membrane type, pore size, and expiration date.
Ordering information: Units with 500 and 1000 ml units include 12 glass fiber prefilters.
Caution: For laboratory use only. Not for in vitro diagnosis or parenterals.
Specifications
- Catalog No:
- 28199-076
- 28199-279
- 28199-268
- 28199-101
- 28199-098
- 28199-280
- 28199-134
- 28199-123
- 28199-155
- 28199-144
- 28199-112
- 28199-177
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)