VWR® Non-Return Valves
Non-Return Valve, Crest 7.8/10.0 mm (5/16/13/32 "), 8/9/10 mm (⁵⁄₁₆/¹¹⁄₃₂/¹³⁄₃₂")
Supplier: VWR International
Jump to:
Product Details & Documents
This polypropylene non-return valve protects tubing and vacuum systems from back flow when pressure falls in the system.
- Polypropylene construction
- Protects against back flow
The recommended working pressure range for this valve is 1.0 to 28.4psi.
Spécifications
- Désignation:Non-Return Valve, Crest 7.8/10.0 mm (5/16/13/32 ")
- Nominal O.D.:8/9/10 mm (⁵⁄₁₆/¹¹⁄₃₂/¹³⁄₃₂")