Nalgene® Check Valve, High-Density Polyethylene, Thermo Scientific
# de catalogue 65551-000
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:Check Valve
- Cat. no.:65551-000
- Supplier No.:6120-0010
Spécifications
A propos de cet article
For use in vacuum lines and to prevent back pressure in filter flasks
Not for use with liquids. Tubing connections are 3/4" long and fit 1/4–5/16" I.D. tubing. Withstands temperatures up to 82.2°C (180°F). Works best between 254–508mm Hg (10–20in. Hg).
Customers Who Bought This Also Bought