KIMBLE® HI-VAC® Valves, with PTFE Plug, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
For vacuum applications up to 5x10-7torr
The shaft is non-rotating and the primary seal is made with a Viton® O-ring on a tapered seat. PTFE shaft can be used continuously at system temperatures of 230°C (446°F) or intermittently at 275°C (527°F).
Designed with a precision external thread for fine control. Valve can be used as a bleed up to 1/2 turn. Full open is at 11/2 turns.
Customers Who Bought This Also Bought