Micro Tissue Grinder with Cap, Wheaton | Avantor
Micro Tissue Grinder with Cap, Wheaton
Micro Tissue Grinder With Cap, 0.5 ml, 115 mm, 13O.D. x 37L mm
Catalog # 14231-334
Fournisseur #:  357535
Micro Tissue Grinder with Cap, Wheaton
Catalog # 14231-334
Micro Tissue Grinder with Cap, Wheaton
Micro Tissue Grinder with Cap, Wheaton
Micro Tissue Grinder With Cap, 0.5 ml, 115 mm, 13O.D. x 37L mm
Price
$429.25
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 2
Product Details & Documents
Grinders for preparation of soft tissue homogenates.
  • Grinding chamber is made of low extractable borosilicate glass
  • Chamber clearance is 0.051–0.152 mm (0.002–0.006")
  • Fluoropolymer resin pestle has 6.4 mm (1/4") diameter stainless steel shaft
  • The open top cap features a fluoropolymer resin-faced silicone rubber liner and a screw thread size of 13-425
Certifications: Glass meets ASTM Type 1 Class A and USP Type 1 standards.
Spécifications
  • Capacité:
    0.5 ml
  • Désignation:
    Micro Tissue Grinder With Cap
  • Overall Length:
    115 mm
  • Chamber Dimensions:
    13O.D. x 37L mm
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Pestle
    Replacement Pestle
    14231-350
    Log in pour la disponibilité
    Case of 2 - $167.44