Tenbroeck Tissue Grinders, Wheaton
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
For homogenizing delicate tissues such as intestines, liver, and heart
Interchangeable pestles and tubes are made of Wheaton 33® low extractable borosilicate glass. Clearance between pestle and grinding chamber is 0.09–0.16mm (0.0035–0.0065"). Glass meets ASTM Type 1 Class A and USP Type 1 standards.
Designed for hand operation, grinders can also be slow-speed motor driven by inserting a rubber stopper (size 00) and metal rod into the pestle. Hollow handle permits packing with ice. Grinders also feature an expanded reservoir and pouring lip.
Customers Who Bought This Also Bought