A propos de cet article
Designed to allow for the development and growth of cell culture from research to production quantities without having to change equipment and revalidate
Each deck holds five bottles. The roller apparatus offers the ability to start with as few as one deck (5 positions for bottles) and add up to 10 decks for a total of 55 bottle positions.
The Modular R2P™ systems feature a larger spacing between the decks than the standard R2P™ systems for inserting and removing roller bottles more easily without disturbing cells adhering to bottle walls.
Accommodates glass or plastic bottles 112 to 121mm in diameter and up to 550mm long. Standard features include an all-position drive system; large digital display and keypad for easy input and monitoring of the settings; programmable soft start and stop with fixed or reversible rotation direction; ten memory locations for setting storage; and built-in monitoring with visual and audible alarms.
Spécifications
- Catalog No:
- 14231-452
- 74840-072
- 74840-074
- 14231-450
- Bottle speed:
- 0.25 - 8.75 rpm (110 mm dia. bottle)
- 0.25 - 8.75 rpm (110 mm dia. bottle)
- 0.25 - 8.75 rpm (110 mm dia. bottle)
- 0.25 - 8.75 rpm (110 mm dia. bottle)
- Electrical:
- 100–120V, 50/60Hz, 35W
- 100–120V, 50/60Hz, 35W
- 100–120V, 50/60Hz, 35W
- 100–120V, 50/60Hz, 35W
- Spacing between decks:
- Standard R2P: 15.2 cm (6"), Modular R2P: 18.1 cm (71/8")
- Standard R2P: 15.2 cm (6"), Modular R2P: 18.1 cm (71/8")
- Standard R2P: 15.2 cm (6"), Modular R2P: 18.1 cm (71/8")
- Standard R2P: 15.2 cm (6"), Modular R2P: 18.1 cm (71/8")
- Dimensions:
- 78.7W x 62.2D x 34.3H cm (31 x 241/2 x 131/2") for drive unit with one deck
- 78.7W x 62.2D x 34.3H cm (31 x 241/2 x 131/2") for drive unit with one deck
- 78.7W x 62.2D x 34.3H cm (31 x 241/2 x 131/2") for drive unit with one deck
- 78.7W x 62.2D x 34.3H cm (31 x 241/2 x 131/2") for drive unit with one deck
- Weight:
- Drive Unit with One Deck: 24 kg (53 lbs.), Each Additional Deck: 7.5 kg (16.5 lbs.)
- Drive Unit with One Deck: 24 kg (53 lbs.), Each Additional Deck: 7.5 kg (16.5 lbs.)
- Drive Unit with One Deck: 24 kg (53 lbs.), Each Additional Deck: 7.5 kg (16.5 lbs.)
- Drive Unit with One Deck: 24 kg (53 lbs.), Each Additional Deck: 7.5 kg (16.5 lbs.)