Thermoplastic Duct Reducers, Labconco®
Fournisseur: Labconco
Certificats
A propos de cet article
PVC coupling type reducers are designed for connecting thermoplastic duct of different diameters
Compare your blower inlet size with your duct size to see if one is necessary.
Customers Who Bought This Also Bought