Accessories for Protector® Stainless Steel Perchloric Acid Laboratory Hoods, Labconco
Supplier: Labconco
Certificates
About this item
Fabricated from vacuum formed unplasticized Type 1 polyvinyl chloride (PVC) and made specifically to handle extremely corrosive atmospheres, these blowers provide optimum chemical resistance to high order oxidizers such as perchloric acid. PVC blowers have a dry powder epoxy-coated base and weather cover for complete protection in extreme weather. Impellers are made of injection molded PVDF to resist highly corrosive atmospheres.
Customers Who Bought This Also Bought