Three-Speed Commercial Blender, 4 L, Waring
# de catalogue 14232-968
Fournisseur: Waring Products
Spécifications
- Cat. no.:14232-968
- Electrical:120 V, 60 Hz, 15 A
- Supplier No.:LBC15
Spécifications
A propos de cet article
Designed for high speed blending of large quantities of highly liquid material.
Stainless steel container has two handles, a lid, and stainless steel blades. Epoxy finished die-cast base houses a 3.75hp motor with a sealed ball bearing system to extend service life. Control panel includes a membrane keypad with Off, Low, Medium, High, and Pulse settings. Non-skid feet help provide stability. Motor speeds: 15,800rpm, 18,000rpm, and 20,800rpm. Supplied with a 2.1m (7') 3-wire cord.
Ordering information: The 120V blender comes with a stainless steel lid while the 240V version comes with a polycarbonate lid.
Customers Who Bought This Also Bought