Pulverizer, Waring
# de catalogue 58983-093
Fournisseur: Waring Products
Spécifications
- Description:Pulverizer, Waring®
- Cat. no.:58983-093
- Supplier No.:SS110
Spécifications
A propos de cet article
Small stainless steel container with special blade configurations reduces small bones, seeds, grains, nuts, soybeans, tablets, soils and similar materials in seconds
Fits all Waring® laboratory blenders (requires an adapter for 2 and 4 liter blenders). Chrome plated die-cast base. Dry capacity: 75g.
Recommended as a replacement for retail coffee mills, which have a short life span in laboratory applications.
Ordering information: Supplied complete with snap tight lid and stainless steel pulverizing blade assembly.
Customers Who Bought This Also Bought