Dry Blending Container, Waring
# de catalogue 58983-100
Fournisseur: Waring Products
Spécifications
- Description:Dry Blending Container, Waring®
- Cat. no.:58983-100
- Supplier No.:SS115
Spécifications
A propos de cet article
Unique blade configuration designed for pulverizing and homogenizing dry substances
Note: Blending such as soil processing requires the use of a larger motor unit.
One liter stainless steel container and lid. Fits 1L Waring® blenders.
Customers Who Bought This Also Bought