Stainless Steel Container for Waring® Blenders, Eberbach
# de catalogue 58984-030
Fournisseur: Eberbach
Spécifications
- Description:Stainless Steel Container for Waring® Blenders, Eberbach
- Cat. no.:58984-030
- Supplier No.:8520
Spécifications
A propos de cet article
Special purpose container assembly with stainless steel screw cover for sealing in contents and 15.9mm (5/8") threaded sampling plug for removal or addition of material
Fits all Waring® power units. Capacity: 1L. Complete with O-ring gaskets for screw cover and sampling plug.
Customers Who Bought This Also Bought