Spectra Mesh® Woven Membrane Filters, Spectrum® Laboratories
Fournisseur: Spectrum Laboratories
Certificats
A propos de cet article
Woven filters are constructed of individual strands woven into a mesh screen, and are characterized by precise mesh openings, percent open area, and mesh thickness.
Woven filters are constructed of individual strands woven into a mesh screen, and are characterized by precise mesh openings, percent open area, and mesh thickness.
Five material types of varying physical, chemical, and thermal characteristics are available for use in a wide array of applications. Available in discs or square sheets.
Spécifications
- Catalog No:
- 28498-764
- 28498-760
- 28498-685
- 28498-656
- 28498-182
- 28498-194
- 28498-188
- 28498-164
- 28498-176
- 28498-170
- 28498-200
- 28498-198
- 28498-242
- 28498-338
- 28498-332
- 28498-344
- 25220-070
- 28498-350
- 28498-212
- 28498-218
- 28498-646
- 28498-644
- 28498-654
- 28498-650
- 28170-190
- Adsorption:
- High
- High
- Low
- High
- High
- High
- High
- High
- High
- High
- High
- High
- Low
- Low (inert)
- Low (inert)
- Low (inert)
- High
- Low (inert)
- Low
- Low
- High
- High
- High
- High
- High
- Best resistance:
- Corrosives
- Corrosives
- Alkalis/Organic
- Corrosives
- Corrosives
- Corrosives
- Corrosives
- Corrosives
- Corrosives
- Corrosives
- Corrosives
- Corrosives
- Alkalis/Organic
- Acids/Alkalis/Organics
- Acids/Alkalis/Organics
- Acids/Alkalis/Organics
- Corrosives
- Acids/Alkalis/Organics
- Alkalis/Organic
- Alkalis/Organic
- Corrosives
- Corrosives
- Corrosives
- Corrosives
- Corrosives
- Material:
- Nylon Mesh
- Nylon Mesh
- Polyester Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Polyester Mesh
- PP mesh
- PP mesh
- PP mesh
- Stainless Steel Mesh
- PP mesh
- Polyester Mesh
- Polyester Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- Nylon Mesh
- pH range:
- 3–10
- 3–10
- 3–13
- 3–10
- 3–10
- 3–10
- 3–10
- 3–10
- 3–10
- 3–10
- 3–10
- 3–10
- 3–13
- 2–14
- 2–14
- 2–14
- 4–12
- 2–14
- 3–13
- 3–13
- 3–10
- 3–10
- 3–10
- 3–10
- 3–10
- Strength/durability:
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High (wet)
- High (wet)
- High (wet)
- Very High
- High (wet)
- High
- High
- High
- High
- High
- High
- High
- Thermal stability:
- up to 180°C
- up to 180°C
- up to 140°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 140°C
- up to 130°C
- up to 130°C
- up to 130°C
- up to 180°C
- up to 130°C
- up to 140°C
- up to 140°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 180°C
- up to 180°C
- Water affinity:
- Hydrophilic
- Hydrophilic
- Mildly Hydrophobic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Mildly Hydrophobic
- Hydrophobic
- Hydrophobic
- Hydrophobic
- Hydrophilic
- Hydrophobic
- Mildly Hydrophobic
- Mildly Hydrophobic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Hydrophilic
- Hydrophilic