Ultrafiltration Membrane Disc Filters
Fournisseur: Cytiva
Certificats
A propos de cet article
Replacement membranes for conventional, nondisposable stirred cell systems.
- For conventional systems
- Variety of diameters available
- Non sterile
Use for concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules. Also for bench or pilot separations prior to scale-up. Available in a broad range of membrane diameters to fit various sizes. Omega™ membrane discs provide unmatched high flow rates and recoveries, and can be washed and reused. Non sterile. May be sanitized using 70% ethanol or 200 ppm sodium hypochlorite.
Spécifications
- Catalog No:
- CA29300-166
- CA29300-162
- CA29300-160
- CA29600-152
- CA29300-150
- CA29300-100
- 29300-166
- CA29300-130
- 29300-098
- CA29300-128
- CA29300-124
- 29300-136
- CA29300-122
- 29300-158
- CA29300-120
- CA29600-154
- 29300-122
- CA29300-138
- CA29300-136
- 29300-132
- CA29300-132
- 29300-152
- 29300-138
- 29300-162
- Max. operating temperature:
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- 0...40 °C (32...104 °F)
- pH range:
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14