Whatman™ Grade 32 Glass Fiber Filters, Whatman products (Cytiva)
Fournisseur: Cytiva
Certificats
A propos de cet article
Highly absorbent, binderless, strong grade widely used in cell harvesting, receptor binding assays, radioimmunoassays, and scintillation counting.
Also specified in the collection of biochemical polymers and sampling of biological fluids.
Spécifications
- Catalog No:
- CA28153-041
- Particle retention:
- 1.0 µm
- Flow rate:
- Slow