ContactUsLinkComponent
Whatman™ Grade 32 Glass Fiber Filters, Whatman products (Cytiva)
Whatman™ Grade 32 Glass Fiber Filters, Whatman products (Cytiva)
Fournisseur:  Cytiva
Certificats
Whatman™ Grade 32 Glass Fiber Filters, Whatman products (Cytiva)
Whatman™ Grade 32 Glass Fiber Filters, Whatman products (Cytiva)
Fournisseur:  Cytiva
Certificats
Spécifications
  • Catalog No:
  • CA28153-041
  • Particle retention:
  • 1.0 µm
  • Flow rate:
  • Slow