Glass Microfiber Filter Paper Grade 151, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Ahlstrom Grade 151 is a binder-free microfiber glass filter designed to retain extremely fine particles.
It is recommended for use in the Toxicity Characteristic Leaching Procedure (TCLP) as described in EPA Method 1311.
Spécifications
- Catalog No:
- 47749-582
- 47749-584
- 47749-578
- 47749-580
- 28496-176
- 28496-174
- 38010-838
- 47749-586
- 28496-180
- 47749-588
- 28496-178
- 28496-184
- 28496-182
- Particle retention:
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- 0.7 microns
- Flow rate:
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Load capacity:
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High