Glass Microfiber Filter Paper Grade 141, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Ahlstrom grade 141 is a binder-free microfiber glass filter paper that is an effective pre-filter, offering better filtration speed and a higher loading capacity than an analytical filter paper of similar particle retention.
Spécifications
- Catalog No:
- 38010-725
- 47749-562
- 28496-168
- 47749-574
- 28496-172
- 28496-170
- 47749-576
- 47749-570
- 47749-564
- 47749-566
- 47749-568
- Particle retention:
- 3.1 microns
- 3.1 microns
- 3.1 microns
- 3.1 microns
- 3.1 microns
- 3.1 microns
- 3.1 microns
- 3.1 microns
- 3.1 microns
- 3.1 microns
- 3.1 microns
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High