SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®
Fournisseur: Labconco
Certificats
A propos de cet article
Solid epoxy work surfaces are 3.2cm (11/4") thick, 65.7cm (257/8") deep, and dished to contain spills.
Solid epoxy cupsink cover 97006-430 glues in cupsink when not in use.
Solid epoxy cupsink cover (model 4005201; sold separately) covers cupsink when not in use.
Customers Who Bought This Also Bought