VWR® Quantitative Filter Paper, Grade 454
Supplier: VWR International
Certificates
About this item
Use for retention of zirconium hydroxide, aluminum hydroxide, cobalt sulfide, and other coarse or gelatinous precipitates.
Specifications
- Catalog No:
- 28306-051
- CA28306-153
- CA28306-131
- CA28306-110
- CA28306-095
- CA28306-051
- CA28306-175
- Particle retention:
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- 10 µm
- Porosity:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Flow rate:
- Medium Fast
- Medium Fast
- Medium Fast
- Medium Fast
- Medium Fast
- Medium Fast
- Medium Fast