SP Bel-Art Urbanti® High-Speed Filter Funnels, TPX®, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Unsurpassed for non-vacuum, high-speed filtration, these clear funnels have internal helicoid ribs to increase the filtration speed.
- Made from autoclavable polymethylpentene plastic
- Glass-clear and unaffected by corrosive liquids
- Heat resistant to 177 °C (350 °F)
Vertical outer ribs prevent airlock.
Customers Who Bought This Also Bought