Anti-ZDHHC16 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:1000-1:5000
Type:
Antigen: ZDHHC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77723-676
- 77723-677
- Antigen symbol:
- ZDHHC16
- ZDHHC16
- Antigen synonyms:
- ZDHHC16|EC 2.3.1.225|Zinc finger DHHC domain-containing protein 16|UNQ2570/PRO6258Palmitoyltransferase ZDHHC16|Abl-philin 2|DHHC-16|APH2
- ZDHHC16|EC 2.3.1.225|Zinc finger DHHC domain-containing protein 16|UNQ2570/PRO6258Palmitoyltransferase ZDHHC16|Abl-philin 2|DHHC-16|APH2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969W1
- Q969W1
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Palmitoyltransferase ZDHHC16 protein (288-377AA)
- Recombinant Human Palmitoyltransferase ZDHHC16 protein (288-377AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Frequently Bought Together