Anti-ADAM2 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: ADAM2
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77714-791
- 77714-790
- Antigen symbol:
- ADAM2
- ADAM2
- Antigen synonyms:
- PH-30|CRYN2|Disintegrin and metalloproteinase domain-containing protein 2|ADAM metallopeptidase domain 2|ADAM 2|PH30-beta|Adam2|CT15|CRYN1|PH30|Ph30 beta|PH 30b|A disintegrin and metalloproteinase domain 2|PH 30|ADAM2_HUMAN|Ftnb|Fertilin subunit beta|Cancer/testis antigen 15|AI323749|Fertilin beta
- PH-30|CRYN2|Disintegrin and metalloproteinase domain-containing protein 2|ADAM metallopeptidase domain 2|ADAM 2|PH30-beta|Adam2|CT15|CRYN1|PH30|Ph30 beta|PH 30b|A disintegrin and metalloproteinase domain 2|PH 30|ADAM2_HUMAN|Ftnb|Fertilin subunit beta|Cancer/testis antigen 15|AI323749|Fertilin beta
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99965
- Q99965
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ADAM2
- Fusion protein of Human ADAM2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together