Anti-HIST1H2BC Rabbit Polyclonal Antibody
Catalog # 77687-899
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:HIST1H2BC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Environmentally Preferable:
- ChIP:Yes
- Form:Liquid
- Gene ID:P62807
- Antigen synonyms:H2B/g|H2BC6|HIST1H2BC|H2BFK|H2B/h|Histone H2B.a|HIST1H2BG|HIST1H2BE|Histone H2B.h|Histone H2B.k|Histone H2B.l|Histone H2B.1 A|H2BFH|H2BC7|H2BC8|H2B/l|HIST1H2BIHistone H2B type 1-C/E/F/G/I|Histone H2B.g|HIST1H2BF|H2B/k|H2BFA|H2BFG|H2BFL|H2BC10|H2B/a|H2BC4
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence around site of Crotonyl-Lys (16) derived from Human Histone H2B type 1-C/E/F/G/I
- Purification:Antigen affinity purified
- Cat. no.:77687-899
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:100-1:1000, ICC:1:20-1:200, IF:1:10-1:100
Type:
Antigen: HIST1H2BC
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human