Anti-AP1M1 Rabbit Polyclonal Antibody
Catalogue # 77690-619
Fournisseur: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antigen symbol:AP1M1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9BXS5
- Antigen synonyms:Clathrin assembly protein complex 1 medium chain|Adaptor-related protein complex 1 mu-1 subunit|CLAPM2|Adaptor protein complex AP-1 mu-1 subunit|HA1 47 kDa subunit|Clathrin coat-associated protein AP47|Clathrin coat assembly protein AP47|AP1M1_HUMAN|Golgi adaptor HA1/AP1 adaptin mu-1 subunit|CLTNM|Golgi adaptor AP 1 47 kDa protein|MU 1A|Mu-adaptin 1|AP-mu chain family member mu1A|Clathrin assembly protein complex 1 medium chain 1|Mu1A-adaptin|AP-1 complex subunit mu-1|Ap1m1|AP47|Clathrin assembly protein complex AP1 mu subunit|Clathrin adaptor protein AP47
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human AP-1 complex subunit mu-1 protein (1-160AA)
- Purification:Antigen affinity purified
- Cat. no.:77690-619
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: AP1M1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human