Anti-MINK1 Rabbit Polyclonal Antibody
Catalogue # 77711-485
Fournisseur: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antigen symbol:MINK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q8N4C8
- Antigen synonyms:ZC3|GCK family kinase MiNK|MINK1|YSK2|MEK kinase kinase 6|Mitogen-activated protein kinase kinase kinase kinase 6|Misshapen-like kinase 1|EC 2.7.11.1|Misshapen/NIK-related kinase|MAP4K6|MEKKK 6|B55|MINK|MAPK/ERK kinase kinase kinase 6
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human MAP4K6
- Purification:Antigen affinity purified
- Cat. no.:77711-485
- Packaging:Vial
Spécifications
À propos de cet article
WB:1:500-1:3000
Type:
Antigen: MINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse