Terfu Fume Extractor Arms, Movex
Fournisseur: Movex
Certificats
A propos de cet article
Fume extractor arms with friction joints are ideal for laboratories and electronics applications
Fume extractor arms with friction joints are ideal for laboratories and electronics applications
Flexible arm allows one-handed adjustments and stays in position when adjusted. Smooth, anodized aluminum tubes and easily dismantled plastic joints facilitate cleaning. Can be mounted on a countertop, ceiling, or wall. Color: white.
Ordering information: For fume extractor arms and hoods with polypropylene and ESD components, contact your Avantor sales representative.
Customers Who Bought This Also Bought