REDISHIP Air Alert 300 Face Velocity Airflow Alarm, Kewaunee®
Catalogue # VFX-0185-28
Fournisseur: Kewaunee
Product available on GSA Advantage®
Spécifications
- Description:Velocity Airflow Alarm
- Cat. no.:VFX-0185-28
- Supplier No.:X-0185-30
Spécifications
À propos de cet article
Air Alert 300 Face Velocity Airflow Alarm consists of a thermistor sensor mounted through the end wall of the hood, and a control monitor that gives both a visual and an audible alarm.
- Thermistor sensor and control monitor
- Visual and audible alarms
The alarm monitors fume hood face velocity and sounds the alarm when the airflow falls below safe levels
The Air Alert 300 operates on a 9 volt DC circuit and comes complete with an adapter that can be plugged into any 1290 VAC receptacle.
A glowing green light signals when conditions are safe again. The control monitor, which is mounted on the hood fascia, also contains a test/reset button that allows the hood user to verify alarm readiness.
Ordering information: REDISHIP units can be shipped within 48 hours of receiving your order. Air Alert 300 is not recommended for use with Auxiliary Air Fume Hoods, and is not for use with Supreme Air LV High Performance Fume Hoods.