REDISHIP Supreme Air® Fume Hood Worktops, Kewaunee®
Fournisseur: Kewaunee
Certificats
A propos de cet article
These modified epoxy resin worktops are available with or without a 5.1cm (2") diameter vent hole
Use the vent hole for vented acid storage base cabinets. The 1.8m (6') and 2.4m (8') models require two base cabinets (vent is provided only for the right-side cabinet). The 2.4m (8') worktop comes in two pieces
Ordering information: REDISHIP units can be shipped within 48 hours of receiving your order.
Customers Who Bought This Also Bought