REDISHIP Supreme Air® Fittings for KMER-Lined Bench and Walk-In Hoods, Kewaunee®
Fournisseur: VWR International
Certificats
A propos de cet article
These remote control fittings consist of a brass angle valve with replaceable seat, a nylon color-coded panel flange with angle serrated hose connector (except VFT-0864-3WV, which features a gooseneck), a remote control rod assembly with a plastic color-coded four-arm handle, black plastic angle flange, and a tank nipple, locknut, and washer
Fittings may be used on either side of the hood. Not for use on Supreme Air Isotope hoods.
Ordering information: REDISHIP units can be shipped within 48 hours of receiving your order.
Customers Who Bought This Also Bought