XPert™ Easy-Order Balance Enclosure Systems, Labconco®
Supplier: Labconco
Certificates
About this item
These systems include an XPert balance enclosure with a Guardian™ Jr
For detailed information about XPert balance enclosures and additional accessories, see 89004-458 series.
For detailed information about FilterMate portable exhausters and filters, see 82010-632 series.
or Guardian™ 1000 digital airflow monitor, a FilterMate™ portable exhauster with HEPA filter, a black solid epoxy dished work surface, an upper exhaust transition adapter, and a 12.7cm (5") O.D. flexible polypropylene hose and clamps.
Customers Who Bought This Also Bought