Nalgene® Closed-Dome Bio Tank Closure with Mixer Support, Thermo Scientific
# de catalogue 66463-210
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:Closed-Dome Bio Tank Closure with Mixer Support
- Cat. no.:66463-210
- Supplier No.:2651-0200
Spécifications
A propos de cet article
Closure with overhead mixer support for use with Closed-Dome Bio Tanks (see 66463-206 series)
Sanitary flange allows aseptic mixing in a closed system. The assembly consists of a 185mm polypropylene screw closure with a 2" sanitary fitting ferrule welded in the center, a 2" silicone gasket, and a true union fitting. Can be connected to other 2" sanitary fittings for drain lines and closed system filling. Individually packaged. Autoclavable, but must be kept vertical if assembled with mixer
Customers Who Bought This Also Bought