StainFrame Slide Staining System, Azer Scientific
# de catalogue 95039-198
Fournisseur: Azer Scientific Inc.
Spécifications
- Dimensions:61L x 10.2W x 5H cm (24 x 4 x 2")
- Description:Slide Staining System
- Cat. no.:95039-198
- Supplier No.:MD-01
Spécifications
A propos de cet article
The StainFrame system is an easy-to-use, multifunctional, and lightweight tool that allows up to fourteen slides to be processed simultaneously.
- Strong ABS Plastic
- Locking Slides
- Multiple-Use Device
- Up to Fourteen Slides Processed Concurrently
The device can be used for multiple procedures such as transport, staining, rinsing, and decolorizing
The system saves time, minimizes stain and reagent use, and yields consistent results. The device is made of durable ABS plastic.
Slides simply lock into place, allowing them to be handled without the threat of breakage.
Customers Who Bought This Also Bought