Flexible Vent Assemblies, Scientific Plastics
Fournisseur: Scientific Plastics
Certificats
A propos de cet article
Venting systems are designed for use in acid and solvent cabinets.
- All vents are manufactured from acid-resistant polyolefin material
- Non-squeezed top
The flexible tubing design allows systems to be installed around potential obstacles within existing cabinet structures. A 3 m (10') length of accessory tubing is available separately for field modifications.
Customers Who Bought This Also Bought