Compact Roller System for Small Bottles, Wheaton®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Compact unit is ideal for benchtop use
Designed to roll vessels that are 108–121mm in diameter and up to 290mm long. Two outside rollers move inward to fit vessels with diameters as small as 75mm. The single-deck unit accommodates two bottles. Use in CO2 environments may necessitate more frequent replacement of motor brushes. Up to two additional deck kits can be added to accommodate an additional two bottles per kit.
Certifications: UL listed. CSA certified. CE marked.
BodyTextAccessories: Additional deck kit for small bottle bench-top roller rack allows for double or triple capacity.
Ordering information: Supplied without bottles. Deck kit consists of assembled roller deck, four support posts, drive belt, necessary hardware, and assembly instructions.
Spécifications
- Catalog No:
- 14231-392
- 14231-390
- Bottle speed:
- 0.1 - 3.8 rpm (110 mm dia. bottle)
- Electrical:
- 100–120V, 50/60Hz, 14W
- Max. ambient temperature:
- 40°C (104°F)
- Shipping weight:
- 8 kg (18 lbs.)
- Dimensions:
- 31W x 32D x 18H cm (121/4 x 123/4 x 71/8")