ES Bench Scales, Ohaus®
Fournisseur: Ohaus
Certificats
A propos de cet article
Rugged, digital scales weigh samples or packages with simple, two-button operation
Scales feature a separate indicator with an easy-to-read 2.54cm (1") high LCD display and a unique three-way mounting bracket that can mount the indicator to the scale base, above the scale platform, or on any vertical surface. Each model comes factory set and calibrated and includes four user-selectable weighing units: kg, lb., oz, and lb:oz. Each scale features an error indicator for over- and under-loads, a low battery indicator, stability indicator, and battery-saving auto shut-off timer (after five minutes of inactivity). Batter power typically provides up to 120 hours/30 hours continuous operation.
Ordering information: Manufacturer’s one-year warranty on parts and labor. Supplied complete with 9V DC/AC adapter and three-way mounting bracket (mounts to base, above base, or to wall). Also available in 220V version; contact your Avantor sales representative.
Spécifications
- Catalog No:
- 46000-418
- Operating temperature range:
- –10 to 40°C (14 to 104°F)
- Voltage:
- 120V