Laboratory Pumps, Oil-Less Reciprocating, Gast®
# de catalogue 54907-086
Fournisseur: Gast Manufacturing
Spécifications
- Description:Oil-Less Reciprocating Pump
- Cat. no.:54907-086
- Supplier No.:1HAB25M100X
Spécifications
A propos de cet article
Special purpose unit functions as both vacuum pump and compressor
Features include oil-less operation, low maintenance and rugged construction. The motor is a single phase, 1/6 hp that delivers 1725rpm.
Ordering information: Complete with gauges, hose barbs, regulators, cord with on/off switch, and carrying handle.
Customers Who Bought This Also Bought