ContactUsLinkComponent
Aspirating Pipette, Corning®
Aspirating Pipette, Corning®
# de catalogue 89091-050
Fournisseur:  Corning
Product available on GSA Advantage®
Aspirator Starter Set Costar
Aspirating Pipette, Corning®
# de catalogue 89091-050
Fournisseur:  Corning
Product available on GSA Advantage®
Spécifications
  • Description:
    Aspirator
  • Cat. no.:
    89091-050
  • Supplier No.:
    4930
Spécifications
A propos de cet article
This aspirating device is used for the removal of liquids from a variety of lab vessels. It is designed for use with an in-house vacuum system. The aspirator allows precise fingertip control of aspiration and presents a sensible alternative to the use of glass pasteur pipettes for supernatant disposal.
  • Designed for a comfortable fit in either hand
  • Can be used with a variety of collection devices
Designed for use with a vacuum pump or an in-house vacuum system and a suitable collection vessel.
Delivery: Supplied with handpiece, grommet for accessory attachment and single-channel adapter for disposable pipette tips (1 to 200 µl). An 8-channel adapter is available separately.
Customers Who Bought This Also Bought