Welch® Barbed Tubing Connectors
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
Chemical-resistant HDPE for use with Welch® Peristaltic Pump Systems (see 84009-696 series)
The barbed fittings kit 84009-610 includes four fittings each of all the tubing connectors for a total of 72 connectors.
The barbed fittings kit 84009-610 includes four fittings each of all the tubing connectors for a total of 72 connectors.
No tools are required to connect tubing connectors to pumps.
Customers Who Bought This Also Bought