VWR® Vacuum Pump Oil No.19
Supplier: VWR International
Certificates
About this item
This is a superior oil based upon special crude oil stocks noted for their durability and fine lubricating qualities.
- Durable
- Fine lubricating qualities
- Clean oil runs coolly in mechanical pumps
- Improves system lifetime
The oil is further enhanced through precision distilling, resulting in a cleaner oil that runs cooler in most mechanical pumps. Cooler operation reduces thermal cracking for longer oil life. Good forepressures are prolonged, reducing backstreaming and there are often noticeable improvements in pumping cycle times.
Customers Who Bought This Also Bought