Female Pipe Adapters, Welch®, Gardner Denver
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
Female pipe adapters are made of stainless steel
It is compatible with Welch® thermocouple vacuum gauges 55008-186 and 55008-189 (see 55008-180 series) and other gauges.
Customers Who Bought This Also Bought