Soldering Tips for PS-90 SensaTemp® Production Soldering Iron, PACE®
Fournisseur: Pace
Certificats
A propos de cet article
Additional tips for PS-90 SensaTemp® Production Soldering Iron (30138-506)
SMT chip component tips remove resistor and capacity chip components; TSOP removal tips are ideal for thin, small outline package type components; PLCC removal tips remove PLCC sockets; SOT-23 removal tips remove small outline transistor type components; and PQFP removal tips remove PQFP type components.
Tips can also be used with PS-80, SP-2A, and IR-70 PACE® irons.
Tips feature a 0.48mm (3/16") shank diameter.
Customers Who Bought This Also Bought