PS-90 SensaTemp® Production Soldering Iron, PACE®
# de catalogue 30138-506
Fournisseur: Pace
Spécifications
- Description:Production Soldering Iron
- Cat. no.:30138-506
- Supplier No.:6993-0199-P1
Spécifications
A propos de cet article
For heavy-duty soldering and general-purpose and SMT rework operations
Provides high thermal capacity and flexibility. Handpiece features PermaGround™ tip grounding system that ensures component safety.
Ordering information: Comes with tip and tool stand. For soldering tips, see 30138-116.
Customers Who Bought This Also Bought