SensaTemp® Handpieces, PACE®
Fournisseur: Pace
Certificats
A propos de cet article
For use with PACE® SensaTemp® compatible stations
SensaTemp® handpieces have a tip temperature range of 204 to 482°C (400 to 900°F) with a temperature stability of ±1.1°C (2°F) at idle from set tip temperature. Choose from the TP-65 ThermoPik™ SMD removal handpiece with vacuum pick-up, the TJ-70 ThermoJet® Hot Air SMD handpiece, and the TT-65 ThermoTweez® SMD removal handpiece.
Ordering information: Handpieces and tips sold separately. Vacuum cup kit 30138-360 includes one small, one medium, and one large cup.
Customers Who Bought This Also Bought