A propos de cet article
Stations offer high capacity, low temperature SMT/thru-hole soldering and desoldering
MBT 250-SDPT Station comes complete with SX-80 Sodr-X-Tractor™ desoldering handpiece, TP-65 ThermoPik™ handpiece with integral vacuum pickup, PS-90 heavy-duty soldering iron, and TT-65 ThermoTweez® handpiece. Also included are accessory kit, VisiFilter® filter, tip and tool holders, tip maintenance station, and instruction manual.
MBT 250-SD Station comes with the SX-80 Sodr-X-Tractor™ desoldering handpiece and the PS-90 heavy-duty soldering iron. System also includes iron holders, VisiFilter®, tip cleaning sponge, accessory kit, and instruction manual.
Three independently controlled channels allow three different SensaTemp handpieces and tips to be operated simultaneously at different temperatures. Digital readout displays the current channel's temperature information, error codes, and programming status. Other features include microprocessor-controlled power supply, auto tip temperature offset compensation, auto-off, and Auto Snap-Vac™ with a 1.2 second minimum vacuum “on time” to prevent clogging. Setback feature automatically reduces tip temperature. All settings, offsets, ranges, and calibrations are kept in memory.
Spécifications
- Catalog No:
- 30138-372
- 30138-370
- H×W×D:
- 13.5 x 16.5 x 26 cm (55/16 x 61/2 x 91/4")
- 13.5 x 16.5 x 26 cm (55/16 x 61/2 x 91/4")
- Weight:
- 5 kg (10 lbs.)
- 5 kg (10 lbs.)